Tisk…

Heptafluoroisopropyl-2-iodotetrafluoroethyl ether 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Iodotetrafluoroethyl heptafluoroisopropyl ether

PC0308-1G PC0308-5G PC0308-25G
APOSPC0308-1GEA 2280 CZK
APOSPC0308-1G APOSPC0308-5G APOSPC0308-25G
Heptafluoroisopropyl-2-iodotetrafluoroethyl ether 97%
Heptafluoroisopropyl-2-iodotetrafluoroethyl ether
Vzorec: C₅F₁₁IO
MW: 411.94 g/mol
Bod varu: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD01320700
CAS číslo: 16005-38-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR