2-Amino-4-(trifluoromethyl)pyrimidine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0306-1G
PC0306-25G
PC0306-5G
APOSPC0306-1GEA
417
CZK
APOSPC0306-1G
APOSPC0306-25G
APOSPC0306-5G
2-Amino-4-(trifluoromethyl)pyrimidine 97%
2-Amino-4-(trifluoromethyl)pyrimidine
Vzorec:
C₅H₄F₃N₃ MW: 163,1 g/mol Bod tání: 174…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00828624 CAS číslo: 16075-42-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...