Tisk…

4-Isocyanato-1-(trifluoroacetyl)piperidine 95+%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Trifluoroacetyl)piperidin-4-yl isocyanate

PC0303-5G PC0303-1G PC0303-25G
APOSPC0303-5GEA 21300 CZK
APOSPC0303-5G APOSPC0303-1G APOSPC0303-25G
4-Isocyanato-1-(trifluoroacetyl)piperidine 95+%
4-Isocyanato-1-(trifluoroacetyl)piperidine

  • May form small amount of sediment.

Vzorec: C₈H₉F₃N₂O₂
MW: 222,17 g/mol
Bod varu: 97…99 °C/0,5mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD03424490
CAS číslo: 493036-44-5
UN: 2206
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR