Tisk…

(2,6-Difluor-3-methylphenyl)acetic acid 98%

Dodavatel: Apollo Scientific
Varování

PC0262-1G PC0262-5G
APOSPC0262-1GEA 2570 CZK
APOSPC0262-1G APOSPC0262-5G
(2,6-Difluor-3-methylphenyl)acetic acid 98%
(2,6-Difluor-3-methylphenyl)acetic acid
Vzorec: C₉H₈F₂O₂
MW: 186,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631338
CAS číslo: 261943-97-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR