2-Fluoro-L-phenylalanine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
H-Phe(2-F)-OH
PC0253-25G
PC0253-5G
APOSPC0253-25GEA
14720
CZK
APOSPC0253-25G
APOSPC0253-5G
2-Fluoro-L-phenylalanine
2-Fluoro-L-phenylalanine
Vzorec:
C₉H₁₀FNO₂ MW: 183,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066448 CAS číslo: 19883-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...