2,3-Difluoro-4-methylbenzylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0251-10G
PC0251-5G
PC0251-1G
APOSPC0251-10GEA
13330
CZK
APOSPC0251-10G
APOSPC0251-5G
APOSPC0251-1G
2,3-Difluoro-4-methylbenzylamine 98%
2,3-Difluoro-4-methylbenzylamine
Vzorec:
C₈H₉F₂N MW: 157,16 g/mol Bod varu: 42 °C/1mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01631409 CAS číslo: 261763-41-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...