2,3-Difluoro-4-methylbenzyl alcohol 98%
Dodavatel: Apollo Scientific
PC0249-10G
PC0249-5G
PC0249-25G
APOSPC0249-10GEA
4860
CZK
APOSPC0249-10G
APOSPC0249-5G
APOSPC0249-25G
2,3-Difluoro-4-methylbenzyl alcohol 98%
2,3-Difluoro-4-methylbenzyl alcohol
Vzorec:
C₈H₈F₂O Storage Temperature: Ambient |
MDL Number:
MFCD01631646 CAS číslo: 252004-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...