Tisk…

2,6-Difluoro-3-toluoyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC0248-1G PC0248-500MG
APOSPC0248-1GEA 1040 CZK
APOSPC0248-1G APOSPC0248-500MG
2,6-Difluoro-3-toluoyl chloride
2,6-Difluoro-3-toluoyl chloride
Vzorec: C₈H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD01631334
CAS číslo: 261763-39-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR