Tisk…

2,6-Difluoro-m-toluic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,6-Difluoro-3-methylbenzoic acid

PC0246-25G PC0246-5G PC0246-1G
APOSPC0246-25GEA 9160 CZK
APOSPC0246-25G APOSPC0246-5G APOSPC0246-1G
2,6-Difluoro-m-toluic acid 97%
2,6-Difluoro-m-toluic acid
Vzorec: C₈H₆F₂O₂
MW: 172,13 g/mol
Bod tání: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD01631332
CAS číslo: 32890-88-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR