2,3,6-Trifluorobenzylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0243-1G
PC0243-5G
APOSPC0243-1GEA
2070
CZK
APOSPC0243-1G
APOSPC0243-5G
2,3,6-Trifluorobenzylamine 98%
2,3,6-Trifluorobenzylamine
Vzorec:
C₇H₆F₃N MW: 161,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236315 CAS číslo: 230295-09-7 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...