3-(Trifluoromethoxy)phenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC0240-25G
PC0240-5G
APOSPC0240-25GEA
2480
CZK
APOSPC0240-25G
APOSPC0240-5G
3-(Trifluoromethoxy)phenylboronic acid 98%
3-(Trifluoromethoxy)phenylboronic acid
Vzorec:
C₇H₆BF₃O₃ Bod tání: 80…89 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01320697 CAS číslo: 179113-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...