Tisk…

α,α,α,3,4-Pentafluorotoluene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,4-Difluorobenzotrifluoride, 1,2-Difluoro-4-(trifluoromethyl)benzene

PC0225-100G PC0225-25G PC0225-5G
APOSPC0225-100GEA 4370 CZK
APOSPC0225-100G APOSPC0225-25G APOSPC0225-5G
α,α,α,3,4-Pentafluorotoluene 98%
α,α,α,3,4-Pentafluorotoluene
Vzorec: C₇H₃F₅
MW: 182,09 g/mol
Bod varu: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00077510
CAS číslo: 32137-19-2
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR