Tisk…

2,4-Dichloro-5-fluorobenzyl alcohol 98%

Dodavatel: Apollo Scientific
Varování

PC0211-10G PC0211-5G PC0211-1G
APOSPC0211-10GEA 6620 CZK
APOSPC0211-10G APOSPC0211-5G APOSPC0211-1G
2,4-Dichloro-5-fluorobenzyl alcohol 98%
2,4-Dichloro-5-fluorobenzyl alcohol
Vzorec: C₇H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD01631380
CAS číslo: 219765-56-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR