2-Chloro-5-(trifluoromethyl)phenylacetonitrile 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0202-1G
PC0202-25G
PC0202-5G
APOSPC0202-1GEA
992
CZK
APOSPC0202-1G
APOSPC0202-25G
APOSPC0202-5G
2-Chloro-5-(trifluoromethyl)phenylacetonitrile 95%
2-Chloro-5-(trifluoromethyl)phenylacetonitrile
Vzorec:
C₉H₅ClF₃N Bod varu: 140…142 °C/12mmHg Bod tání: 30…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631472 CAS číslo: 22902-88-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...