Tisk…

α-Bromo-3-chloro-5-fluorotoluene 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC0198-5G PC0198-1G PC0198-25G
APOSPC0198-5GEA 1080 CZK
APOSPC0198-5G APOSPC0198-1G APOSPC0198-25G
α-Bromo-3-chloro-5-fluorotoluene 98%
α-Bromo-3-chloro-5-fluorotoluene
Vzorec: C₇H₅BrClF
Storage Temperature: Ambient
MDL Number: MFCD03788472
CAS číslo: 493024-39-8
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR