Tisk…

5-Chloro-2-(trifluoromethyl)benzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Bromomethyl)-5-chloro-2-(trifluoromethyl)benzene, 2-(Bromomethyl)-4-chloro-1-(trifluoromethyl)benzene

PC0195-10G PC0195-5G PC0195-1G
APOSPC0195-10GEA 3060 CZK
APOSPC0195-10G APOSPC0195-5G APOSPC0195-1G
5-Chloro-2-(trifluoromethyl)benzyl bromide 98%
5-Chloro-2-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₅BrClF₃
MW: 273,48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631356
CAS číslo: 261763-24-0
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR