3-Chloro-2-fluoro-5-(trifluoromethyl)phenol
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0165-25G
PC0165-5G
PC0165-250MG
APOSPC0165-25GEA
10350
CZK
APOSPC0165-25G
APOSPC0165-5G
APOSPC0165-250MG
3-Chloro-2-fluoro-5-(trifluoromethyl)phenol
3-Chloro-2-fluoro-5-(trifluoromethyl)phenol
Vzorec:
C₇H₃ClF₄O MW: 214,55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631514 CAS číslo: 261763-12-6 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...