Tisk…

3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride 98%

Dodavatel: Apollo Scientific

PC0153-5G PC0153-10G PC0153-1G
APOSPC0153-5GEA 2780 CZK
APOSPC0153-5G APOSPC0153-10G APOSPC0153-1G
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride 98%
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride
Vzorec: C₈H₂Cl₂F₄O
MW: 261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631344
CAS číslo: 186517-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR