Tisk…

5-Chloro-4-(trifluoromethyl)pyridin-2-amine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-5-chloro-4-(trifluoromethyl)pyridine

PC01524-1G PC01524-250MG PC01524-5G
APOSPC01524-1GEA 1700 CZK
APOSPC01524-1G APOSPC01524-250MG APOSPC01524-5G
5-Chloro-4-(trifluoromethyl)pyridin-2-amine
5-Chloro-4-(trifluoromethyl)pyridin-2-amine
Vzorec: C6H4ClF3N2
MW: 196,56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18255015
CAS číslo: 1095823-39-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR