Tisk…

4-Bromo-3-fluorothiophenol 95%

Dodavatel: Apollo Scientific
Varování

PC01517-1G PC01517-5G PC01517-250MG
APOSPC01517-1GEA 7870 CZK
APOSPC01517-1G APOSPC01517-5G APOSPC01517-250MG
4-Bromo-3-fluorothiophenol 95%
4-Bromo-3-fluorothiophenol
Vzorec: C₆H₄BrFS
MW: 207.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09475476
CAS číslo: 942473-86-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR