Tisk…

3-Fluoro-2-methoxypyridine 95%

Dodavatel: Apollo Scientific

PC01473-10G PC01473-25G PC01473-5G
APOSPC01473-10GEA 4100 CZK
APOSPC01473-10G APOSPC01473-25G APOSPC01473-5G
3-Fluoro-2-methoxypyridine 95%
3-Fluoro-2-methoxypyridine
Vzorec: C₆H₆FNO
MW: 127.12
Bod vzplanutí: <60 °C
Storage Temperature: Ambient
MDL Number: MFCD03094945
CAS číslo: 884494-69-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR