Tisk…

3-Chloro-2-fluoro-5-(trifluoromethyl)benzamide

Dodavatel: Apollo Scientific
Varování

PC0147-1G PC0147-10G PC0147-5G
APOSPC0147-1GEA 744 CZK
APOSPC0147-1G APOSPC0147-10G APOSPC0147-5G
3-Chloro-2-fluoro-5-(trifluoromethyl)benzamide
3-Chloro-2-fluoro-5-(trifluoromethyl)benzamide
Vzorec: C₈H₄ClF₄NO
Storage Temperature: Ambient
MDL Number: MFCD01631399
CAS číslo: 129931-46-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR