3-Chloro-2-fluoro-5-(trifluoromethyl)anisole 98%
Dodavatel: Apollo Scientific
PC0142-5G
PC0142-1G
PC0142-25G
PC0142-100G
APOSPC0142-5GEA
4860
CZK
APOSPC0142-5G
APOSPC0142-1G
APOSPC0142-25G
APOSPC0142-100G
3-Chloro-2-fluoro-5-(trifluoromethyl)anisole 98%
3-Chloro-2-fluoro-5-(trifluoromethyl)anisole
Vzorec:
C₈H₅ClF₄O Storage Temperature: Ambient |
MDL Number:
MFCD01631517 CAS číslo: 261763-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...