5-Chloro-2-fluorophenylacetonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC0138-10G
PC0138-5G
PC0138-25G
APOSPC0138-10GEA
827
CZK
APOSPC0138-10G
APOSPC0138-5G
APOSPC0138-25G
5-Chloro-2-fluorophenylacetonitrile 97%
5-Chloro-2-fluorophenylacetonitrile
Vzorec:
C₈H₅ClFN Storage Temperature: Ambient |
MDL Number:
MFCD01631438 CAS číslo: 75279-54-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...