6-Chloro-2-fluoro-m-cresol 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
6-Chloro-2-fluoro-3-methylphenol
PC0119-25G
PC0119-5G
PC0119-100G
APOSPC0119-25GEA
14120
CZK
APOSPC0119-25G
APOSPC0119-5G
APOSPC0119-100G
6-Chloro-2-fluoro-m-cresol 98%
6-Chloro-2-fluoro-m-cresol
Vzorec:
C₇H₆ClFO Bod tání: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631376 CAS číslo: 261762-91-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...