2-Chloro-6-fluoro-m-cresol
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-6-fluoro-3-methylphenol, 2-Choro-6-fluoro-3-methylphenol
PC0118-1G
PC0118-5G
APOSPC0118-1GEA
622
CZK
APOSPC0118-1G
APOSPC0118-5G
2-Chloro-6-fluoro-m-cresol
2-Chloro-6-fluoro-m-cresol
Vzorec:
C₇H₆ClFO MW: 160,58 g/mol Bod varu: 211 °C Bod tání: 47…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631366 CAS číslo: 261762-90-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...