Tisk…

2-Chloro-6-fluoro-3-methylbenzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC0112-5G PC0112-1G
APOSPC0112-5GEA 7180 CZK
APOSPC0112-5G APOSPC0112-1G
2-Chloro-6-fluoro-3-methylbenzylamine 98%
2-Chloro-6-fluoro-3-methylbenzylamine
Vzorec: C₈H₉ClFN
Storage Temperature: Ambient
MDL Number: MFCD01631395
CAS číslo: 261762-85-0
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR