Tisk…

2-Chloro-6-fluoro-3-methylbenzamide 98%

Dodavatel: Apollo Scientific

PC0095-5G PC0095-1G
APOSPC0095-5GEA 2780 CZK
APOSPC0095-5G APOSPC0095-1G
2-Chloro-6-fluoro-3-methylbenzamide 98%
2-Chloro-6-fluoro-3-methylbenzamide
Vzorec: C₈H₇ClFNO
Storage Temperature: Ambient
MDL Number: MFCD01631394
CAS číslo: 286474-60-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR