Tisk…

2-Chloro-6-fluoro-3-methylanisole 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC0090-10G PC0090-5G PC0090-1G
APOSPC0090-10GEA 5390 CZK
APOSPC0090-10G APOSPC0090-5G APOSPC0090-1G
2-Chloro-6-fluoro-3-methylanisole 98%
2-Chloro-6-fluoro-3-methylanisole
Vzorec: C₈H₈ClFO
MW: 174,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631367
CAS číslo: 261762-79-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR