6'-Chloro-2'-fluoro-3'-methylacetophenone 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC0089-25G
PC0089-5G
PC0089-1G
APOSPC0089-25GEA
15380
CZK
APOSPC0089-25G
APOSPC0089-5G
APOSPC0089-1G
6'-Chloro-2'-fluoro-3'-methylacetophenone 97%
6'-Chloro-2'-fluoro-3'-methylacetophenone
Vzorec:
C₉H₈ClFO Storage Temperature: Ambient |
MDL Number:
MFCD01631378 CAS číslo: 261762-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...