Tisk…

(Pentafluorophenoxy)acetyl chloride 99%

Dodavatel: Apollo Scientific
Nebezpečí

PC0088-1G PC0088-5G
APOSPC0088-1GEA 1870 CZK
APOSPC0088-1G APOSPC0088-5G
(Pentafluorophenoxy)acetyl chloride 99%
(Pentafluorophenoxy)acetyl chloride
Vzorec: C₈H₂ClF₅O₂
Bod varu: 116…117 °C/20mmHg
Hustota: 1,6 g/cm³ (25 °C)
Bod vzplanutí: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320685
CAS číslo: 55502-53-9
EINECS: 259-676-2
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR