α-Bromo-5-chloro-2-fluorotoluene
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-Chloro-2-fluorobenzyl bromide, 2-(Bromomethyl)-4-chloro-1-fluorobenzene
PC0080-10G
PC0080-5G
PC0080-25G
APOSPC0080-10GEA
827
CZK
APOSPC0080-10G
APOSPC0080-5G
APOSPC0080-25G
α-Bromo-5-chloro-2-fluorotoluene
α-Bromo-5-chloro-2-fluorotoluene
Vzorec:
C₇H₅BrClF MW: 223.47 Bod varu: 44…45 °C/0.5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01631437 CAS číslo: 71916-91-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...