Tisk…

2-Amino-6-(trifluoromethyl)benzothiazole 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 6-(Trifluoromethyl)benzo[d]thiazol-2-amine

PC0072-1G PC0072-5G PC0072-10G
APOSPC0072-1GEA 1040 CZK
APOSPC0072-1G APOSPC0072-5G APOSPC0072-10G
2-Amino-6-(trifluoromethyl)benzothiazole 98%
2-Amino-6-(trifluoromethyl)benzothiazole
Vzorec: C₈H₅F₃N₂S
MW: 218,20 g/mol
Bod tání: 120…124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269597
CAS číslo: 777-12-8
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR