Tisk…

3-Fluoro-4-toluoyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Fluoro-4-methylbenzoyl chloride, 3-Fluoro-4-methylbenzene-1-carbonyl chloride

PC0065-5G PC0065-25G
APOSPC0065-5GEA 1080 CZK
APOSPC0065-5G APOSPC0065-25G
3-Fluoro-4-toluoyl chloride 98%
3-Fluoro-4-toluoyl chloride
Vzorec: C₈H₆ClFO
MW: 172,59 g/mol
Bod varu: 216 °C
Hustota: 1,25 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00671774
CAS číslo: 59189-97-8
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR