Tisk…

5-Chloro-2-fluorobenzyl alcohol 97%

Dodavatel: Apollo Scientific
Varování

PC0063-1G PC0063-5G
APOSPC0063-1GEA 992 CZK
APOSPC0063-1G APOSPC0063-5G
5-Chloro-2-fluorobenzyl alcohol 97%
5-Chloro-2-fluorobenzyl alcohol
Vzorec: C₇H₆ClFO
MW: 160,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631436
CAS číslo: 188723-58-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR