Tisk…

3-Chloro-4-fluorobenzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Chloro-4-fluorobenzyl Alcohol, (3-Chloro-4-fluorophenyl)methanol

PC0061-1G PC0061-25G
APOSPC0061-1GEA 622 CZK
APOSPC0061-1G APOSPC0061-25G
3-Chloro-4-fluorobenzyl alcohol
3-Chloro-4-fluorobenzyl alcohol
Vzorec: C₇H₆ClFO
MW: 160,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631550
CAS číslo: 161446-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR