Tisk…

2-Chloro-5-fluorobenzoyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC0040-1G PC0040-500MG
APOSPC0040-1GEA 0 CZK
APOSPC0040-1G APOSPC0040-500MG
2-Chloro-5-fluorobenzoyl chloride 98%
2-Chloro-5-fluorobenzoyl chloride
Vzorec: C₇H₃Cl₂FO
Bod varu: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD01631417
CAS číslo: 21900-51-6
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR