Tisk…

2-Chloro-5-fluorobenzamide 98%

Dodavatel: Apollo Scientific

PC0029-25G PC0029-5G PC0029-1G
APOSPC0029-25GEA 6620 CZK
APOSPC0029-25G APOSPC0029-5G APOSPC0029-1G
2-Chloro-5-fluorobenzamide 98%
2-Chloro-5-fluorobenzamide
Vzorec: C₇H₅ClFNO
MW: 173,57 g/mol
Bod tání: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD01631465
CAS číslo: 88487-25-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR