Tisk…

3-Chloro-2-fluoroanisole 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Chloro-2-fluoro-3-methoxybenzene

PC0026-1G PC0026-5G PC0026-25G
APOSPC0026-1GEA 622 CZK
APOSPC0026-1G APOSPC0026-5G APOSPC0026-25G
3-Chloro-2-fluoroanisole 97%
3-Chloro-2-fluoroanisole
Vzorec: C₇H₆ClFO
MW: 160.58
Storage Temperature: Ambient
MDL Number: MFCD01631570
CAS číslo: 261762-56-5
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR