(3-Chloro-2,6-difluorophenyl)acetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC0018-1G
PC0018-5G
PC0018-10G
APOSPC0018-1GEA
3110
CZK
APOSPC0018-1G
APOSPC0018-5G
APOSPC0018-10G
(3-Chloro-2,6-difluorophenyl)acetic acid 98%
(3-Chloro-2,6-difluorophenyl)acetic acid
Vzorec:
C₈H₅ClF₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01631328 CAS číslo: 261762-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...