Tisk…

α-Bromo-3-chloro-2,6-difluorotoluene 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC0009-5G PC0009-1G
APOSPC0009-5GEA 3940 CZK
APOSPC0009-5G APOSPC0009-1G
α-Bromo-3-chloro-2,6-difluorotoluene 97%
α-Bromo-3-chloro-2,6-difluorotoluene
Vzorec: C₇H₄BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD01631326
CAS číslo: 261762-47-4
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR