α-Bromo-2-chloro-3,6-difluorotoluene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0008-1G
PC0008-5G
PC0008-25G
APOSPC0008-1GEA
662
CZK
APOSPC0008-1G
APOSPC0008-5G
APOSPC0008-25G
α-Bromo-2-chloro-3,6-difluorotoluene 98%
α-Bromo-2-chloro-3,6-difluorotoluene
Vzorec:
C₇H₄BrClF₂ MW: 241.46 Bod varu: 218 Bod tání: 50 Storage Temperature: Ambient |
MDL Number:
MFCD01631446 CAS číslo: 90292-67-4 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...