Tisk…

2-Chloro-3,6-difluorobenzyl alcohol 98%

Dodavatel: Apollo Scientific

PC0003-5G
APOSPC0003-5GEA 2280 CZK
APOSPC0003-5G
2-Chloro-3,6-difluorobenzyl alcohol 98%
2-Chloro-3,6-difluorobenzyl alcohol
Vzorec: C₇H₅ClF₂O
MW: 178,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631445
CAS číslo: 261762-44-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR