Tisk…

(R)-(-)-2-Aminobutyric acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (R)-()-2-Aminobutyric acid, D-2-Aminobutyric acid, D(-)-2-Aminobutyric acid, H-D-Abu-OH

OR9988-25G OR9988-100G
APOSOR9988-25GEA 622 CZK
APOSOR9988-25G APOSOR9988-100G
(R)-(-)-2-Aminobutyric acid 98%
(R)-(-)-2-Aminobutyric acid
Vzorec: C₄H₉NO₂
MW: 103,12 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064414
CAS číslo: 2623-91-8
EINECS: 220-084-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR