Tisk…

(±)-2-Aminobutyric acid

Dodavatel: Apollo Scientific

Synonyms: H-DL-Abu-OH, DL-2-Aminobutyric acid, DL-α-Amino-N-Butyric Acid

OR9986-100G OR9986-25G OR9986-500G
APOSOR9986-100GEA 1870 CZK
APOSOR9986-100G APOSOR9986-25G APOSOR9986-500G
(±)-2-Aminobutyric acid
(±)-2-Aminobutyric acid
Vzorec: C₄H₉NO₂
MW: 103,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008093
CAS číslo: 2835-81-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR