o-Toluenesulphonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Toluene-o-sulphonyl chloride, Toluene-2-sulphonyl chloride, 2-Toluenesulphonyl chloride
OR9961-1G
OR9961-5G
OR9961-10G
APOSOR9961-1GEA
3310
CZK
APOSOR9961-1G
APOSOR9961-5G
APOSOR9961-10G
o-Toluenesulphonyl chloride 97%
o-Toluenesulphonyl chloride
Vzorec:
C₇H₇ClO₂S MW: 190,65 g/mol Bod varu: 254 °C Bod tání: 10 °C Hustota: 1,32 g/cm³ (25 °C) Bod vzplanutí: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024874 CAS číslo: 133-59-5 EINECS: 205-113-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...