4-Chloro-3-methoxy-2-methylpyridine-N-oxide 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloro-3-methoxy-2-methylpyridine 1-oxide
OR9945-100G
OR9945-25G
OR9945-5G
APOSOR9945-100GEA
13690
CZK
APOSOR9945-100G
APOSOR9945-25G
APOSOR9945-5G
4-Chloro-3-methoxy-2-methylpyridine-N-oxide 98%
4-Chloro-3-methoxy-2-methylpyridine-N-oxide
Vzorec:
C₇H₈ClNO₂ MW: 173,60 g/mol Bod tání: 96…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093995 CAS číslo: 122307-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...