3-Chloro-o-toluic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR9938-100G
OR9938-25G
APOSOR9938-100GEA
2070
CZK
APOSOR9938-100G
APOSOR9938-25G
3-Chloro-o-toluic acid 98%
3-Chloro-o-toluic acid
Vzorec:
C₈H₇ClO₂ MW: 170,6 g/mol Bod tání: 163…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053312 CAS číslo: 7499-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...