Cysteaminium chloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-MEA
OR9920T-100G
OR9920T-25G
APOSOR9920T-100GEA
1740
CZK
APOSOR9920T-100G
APOSOR9920T-25G
Cysteaminium chloride
Cysteaminium chloride
Used as an antioxidant & in radiation therapy.
Vzorec:
C₂H₇NS·HCl Bod tání: 66…68 °C Hustota: 0,75 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012904 CAS číslo: 156-57-0 UN: 3335 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...