Tisk…

4-(1-Azepanyl)aniline 95%

Dodavatel: Apollo Scientific
Varování

OR9910-100MG OR9910-1G OR9910-500MG
APOSOR9910-100MGEA 830 CZK
APOSOR9910-100MG APOSOR9910-1G APOSOR9910-500MG
4-(1-Azepanyl)aniline 95%
4-(1-Azepanyl)aniline
Vzorec: C₁₂H₁₈N₂
MW: 190,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01536453
CAS číslo: 57356-18-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR